Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ints10
Entrez Gene ID
70885
Gene Name
integrator complex subunit 10
Gene Aliases
4921521J11Rik, AI462004
Chromosome Location
Chr.8: 68793928 - 68829410 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293791.1 NP_001280720.1 6 - 7 753 70
NM_001293792.1 NP_001280721.1 6 - 7 753 70
NM_027590.4 NP_081866.1 6 - 7 753 70
GenBank mRNA AK014939.1 - 6 - 7 729 70
AK030974.1 - 6 - 7 717 70
AK034498.1 - 6 - 7 669 70
AK040574.1 - 6 - 7 754 70
AK047093.1 - 6 - 7 748 70
AK147743.1 - 6 - 7 727 70
BC031923.1 - 6 - 7 716 70

View Full Product Details