Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tex10
Entrez Gene ID
269536
Gene Name
testis expressed gene 10
Gene Aliases
2610206N19Rik, 2810462N03Rik, BC006867
Chromosome Location
Chr.4: 48430858 - 48473464 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172304.3 NP_758508.2 12 - 13 2595 95
GenBank mRNA AK132870.1 - 12 - 13 2596 95
AK141288.1 - 11 - 12 2374 95
BC006867.1 - 9 - 10 1842 95
BC151176.1 - 11 - 12 2346 95
BC151177.1 - 11 - 12 2346 95

View Full Product Details