Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akap8
Entrez Gene ID
56399
Gene Name
A kinase (PRKA) anchor protein 8
Gene Aliases
1200016A02Rik, AA673585, AKAP-8, AKAP95, AU015639
Chromosome Location
Chr.17: 32303672 - 32321327 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB028920.1 - 12 - 13 1563 71
AK004801.1 - 12 - 13 1569 71
AK089092.1 - 12 - 13 1576 71
BC042602.1 - 12 - 13 2446 71
BC054433.1 - 10 - 11 2143 71
BC060170.1 - 12 - 13 2443 71
BC060209.1 - 11 - 12 2059 71
BC078448.1 - 11 - 12 1949 71
BC125521.1 - 12 - 13 1559 71
BC137746.1 - 12 - 13 1559 71

View Full Product Details