The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163734.1 | NP_001157206.1 | 1 - 2 | 358 | 78 |
NM_028244.2 | NP_082520.2 | 4 - 5 | 674 | 78 | |
GenBank mRNA | AK129078.1 | - | 1 - 2 | 358 | 78 |
BC016569.1 | - | 4 - 5 | 674 | 78 |