The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163734.1 | NP_001157206.1 | 3 - 4 | 543 | 100 |
NM_028244.2 | NP_082520.2 | 6 - 7 | 859 | 100 | |
GenBank mRNA | AK129078.1 | - | 3 - 4 | 543 | 100 |
BC016569.1 | - | 6 - 7 | 859 | 100 |