Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ephx3
Entrez Gene ID
71932
Gene Name
epoxide hydrolase 3
Gene Aliases
2310063B19Rik, Abhd9
Chromosome Location
Chr.17: 32183770 - 32189850 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033163.3 NP_001028335.1 5 - 6 925 95
GenBank mRNA AK132483.1 - 5 - 6 926 95
BC145797.1 - 5 - 6 990 95

View Full Product Details