The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033163.3 | NP_001028335.1 | 6 - 7 | 1066 | 74 |
GenBank mRNA | AK010021.1 | - | 6 - 7 | 1055 | 74 |
AK132483.1 | - | 6 - 7 | 1067 | 74 | |
BC145797.1 | - | 6 - 7 | 1131 | 74 |