Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trat1
Entrez Gene ID
77647
Gene Name
T cell receptor associated transmembrane adaptor 1
Gene Aliases
C030046M14Rik, Tcrim, Trim
Chromosome Location
Chr.16: 48731914 - 48810520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198297.3 NP_938039.2 4 - 5 254 88
GenBank mRNA AJ297969.1 - 4 - 5 272 88
AK030807.1 - 4 - 5 437 88
AK138735.1 - - 320 88
BC104350.1 - 3 - 4 217 88
BC104351.1 - 4 - 5 254 88

View Full Product Details