Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
4930453H23Rik
Entrez Gene ID
100503481
Gene Name
RIKEN cDNA 4930453H23 gene
Gene Aliases
-
Chromosome Location
Chr.X: 21206580 - 21221672 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252013.1 NP_001238942.1 2 - 3 742 78
GenBank mRNA AK015451.1 - 2 - 3 744 78


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream synovial sarcoma, X member A, breakpoint 1 Ssxa1 Gm1532, Ssxa -97.2kb Anti-sense Chr. - 21115816 - 21122088 on Build GRCm38
Downstream predicted gene 6938 Gm6938 EG629024 92.4kb Sense Chr. - 21312210 - 21334727 on Build GRCm38

View Full Product Details