The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182839.2 | NP_878259.1 | 4 - 4 | 5028 | 81 |
GenBank mRNA | AK013631.1 | - | 1 - 1 | 969 | 81 |
BC054803.1 | - | 4 - 4 | 4968 | 81 | |
BM898784.1 | - | 1 - 1 | 81 |