Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shroom3
Entrez Gene ID
27428
Gene Name
shroom family member 3
Gene Aliases
AL022960, D5Ertd287e, Shrm, Shrm3
Chromosome Location
Chr.5: 92683435 - 92965759 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015756.2 NP_056571.2 2 - 3 552 92
GenBank mRNA AF199421.1 - 2 - 3 552 92
BC145113.1 - 2 - 3 362 92

View Full Product Details