Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shroom3
Entrez Gene ID
27428
Gene Name
shroom family member 3
Gene Aliases
AL022960, D5Ertd287e, Shrm, Shrm3
Chromosome Location
Chr.5: 92683435 - 92965759 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077595.2 NP_001071063.1 2 - 3 308 108
NM_001077596.2 NP_001071064.1 2 - 3 356 108
NM_015756.2 NP_056571.2 3 - 4 680 108
GenBank mRNA AF199421.1 - 3 - 4 680 108
AF199422.1 - 2 - 3 248 108
AK147493.1 - 2 - 3 291 108
AK164918.1 - 2 - 3 267 108
BC145113.1 - 3 - 4 490 108
BY732045.1 - 2 - 3 351 108
CJ045556.1 - 2 - 3 357 108
CJ103792.1 - 2 - 3 313 108

View Full Product Details