The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009818.1 | NP_001009818.1 | 2 - 3 | 259 | 118 |
GenBank mRNA | AK028475.1 | - | 2 - 3 | 191 | 118 |
AK166517.1 | - | 2 - 3 | 280 | 118 | |
AK170945.1 | - | 1 - 2 | 69 | 118 | |
AK171368.1 | - | 2 - 3 | 280 | 118 | |
BC031456.1 | - | 2 - 3 | 166 | 118 | |
BC062206.1 | - | 2 - 3 | 300 | 118 | |
BC064466.1 | - | 2 - 3 | 259 | 118 |