Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rchy1
Entrez Gene ID
68098
Gene Name
ring finger and CHY zinc finger domain containing 1
Gene Aliases
ARNIP, CHIMP, Pirh2, Zfp363, Znf363
Chromosome Location
Chr.5: 91948842 - 91963224 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_073442.1 2 - 3 432 68
GenBank mRNA AK015870.1 - 2 - 3 433 68

View Full Product Details