Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr45
Entrez Gene ID
54636
Gene Name
WD repeat domain 45
Gene Aliases
C79260, DXImx38e, JM5, Sfc19, WIPI-4, Wdrx1
Chromosome Location
Chr.X: 7721995 - 7728201 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290792.1 NP_001277721.1 7 - 8 695 65
NM_001290794.1 NP_001277723.1 7 - 8 591 65
NM_001290795.1 NP_001277724.1 7 - 8 695 65
NM_172372.2 NP_758960.1 7 - 8 691 65
GenBank mRNA AF229635.1 - 7 - 8 669 65
AK145616.1 - 7 - 8 591 65
AK208276.1 - 2 - 3 156 65
BC011479.1 - 7 - 8 662 65

View Full Product Details