The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290536.1 | NP_001277465.1 | 1 - 2 | 884 | 56 |
NM_001290537.1 | NP_001277466.1 | 2 - 3 | 504 | 56 | |
NM_138604.3 | NP_613070.2 | 1 - 2 | 884 | 56 | |
GenBank mRNA | AK010750.1 | - | 2 - 3 | 502 | 56 |
AK149777.1 | - | 2 - 3 | 505 | 56 | |
AK155131.1 | - | 1 - 2 | 860 | 56 | |
AK167226.1 | - | 1 - 2 | 110 | 56 | |
AK169921.1 | - | 2 - 3 | 470 | 56 | |
AK171874.1 | - | 1 - 2 | 625 | 56 | |
BC016529.1 | - | 1 - 2 | 884 | 56 | |
BC051111.1 | - | 1 - 2 | 623 | 56 |