Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Diap2
Entrez Gene ID
54004
Gene Name
diaphanous homolog 2 (Drosophila)
Gene Aliases
Dia3, Diaph2, Drf2, E430022I22Rik
Chromosome Location
Chr.X: 129749668 - 130465834 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK088648.1 - 1 - 2 323 79
AK153986.1 - 1 - 2 377 79

View Full Product Details