The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172493.2 | NP_766081.1 | 3 - 4 | 582 | 106 |
GenBank mRNA | AB713187.1 | - | 3 - 4 | 356 | 106 |
AK088648.1 | - | 2 - 3 | 495 | 106 | |
AK153986.1 | - | 2 - 3 | 549 | 106 | |
AY312280.1 | - | 3 - 4 | 519 | 106 |