Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rufy3
Entrez Gene ID
52822
Gene Name
RUN and FYVE domain containing 3
Gene Aliases
2810428M05Rik, 6330416M07Rik, AW455998, AW538594, D5Bwg0860e, Ripx, Rpipx
Chromosome Location
Chr.5: 88564780 - 88651417 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289774.1 NP_001276703.1 12 - 13 1598 148
GenBank mRNA AK154941.1 - 12 - 13 1599 148
BP765560.1 - 2 - 3 203 148

View Full Product Details