Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rp2h
Entrez Gene ID
19889
Gene Name
retinitis pigmentosa 2 homolog (human)
Gene Aliases
AI662636, Rp2
Chromosome Location
Chr.X: 20364481 - 20405653 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_110966.1 4 - 5 1213 82
GenBank mRNA AK044002.1 - 4 - 5 1120 82

View Full Product Details