The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290430.1 | NP_001277359.1 | 1 - 2 | 237 | 75 |
NM_133991.2 | NP_598752.2 | 1 - 2 | 237 | 75 | |
GenBank mRNA | AK004357.1 | - | 1 - 2 | 195 | 75 |
AK036313.1 | - | 1 - 2 | 195 | 75 | |
BC011144.1 | - | 1 - 2 | 141 | 75 | |
BC034880.1 | - | 1 - 2 | 107 | 75 | |
BY099531.1 | - | 1 - 2 | 238 | 75 |