Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pink1
Entrez Gene ID
68943
Gene Name
PTEN induced putative kinase 1
Gene Aliases
1190006F07Rik, AU042772, AW557854, BRPK, mFLJ00387
Chromosome Location
Chr.4: 138313409 - 138326296 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026880.2 NP_081156.2 4 - 5 1031 73
GenBank mRNA AB053476.1 - 4 - 5 963 73
AK050322.1 - 4 - 5 1011 73
AK155474.1 - 4 - 5 1017 73
AK165480.1 - 4 - 5 1010 73
AK180089.1 - 1 - 2 92 73
BC018594.1 - 4 - 5 994 73
BC067066.1 - 4 - 5 999 73

View Full Product Details