Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pqlc2
Entrez Gene ID
212555
Gene Name
PQ loop repeat containing 2
Gene Aliases
BC019216
Chromosome Location
Chr.4: 139294030 - 139310711 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145384.2 NP_663359.2 1 - 2 70 65
GenBank mRNA AK081661.1 - 1 - 2 57 65
AK156668.1 - 1 - 2 70 65
BB844529.1 - 1 - 2 70 65

View Full Product Details