The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007711.3 | NP_031737.1 | 3 - 4 | 795 | 79 |
NM_173873.1 | NP_776298.1 | 4 - 5 | 971 | 79 | |
NM_173874.1 | NP_776299.1 | 4 - 5 | 971 | 79 | |
NM_173876.3 | NP_776301.1 | 3 - 4 | 516 | 79 | |
GenBank mRNA | AF029347.1 | - | 3 - 4 | 419 | 79 |
AK141136.1 | - | 4 - 5 | 909 | 79 | |
AK147364.1 | - | 3 - 4 | 951 | 79 | |
AK147548.1 | - | 3 - 4 | 953 | 79 | |
AK147621.1 | - | 3 - 4 | 953 | 79 | |
AK149276.1 | - | 4 - 5 | 510 | 79 | |
AK150688.1 | - | 4 - 5 | 900 | 79 | |
AK159698.1 | - | 4 - 5 | 945 | 79 | |
AK161327.1 | - | 4 - 5 | 899 | 79 | |
AK162639.1 | - | 4 - 5 | 912 | 79 | |
AK163732.1 | - | 4 - 5 | 904 | 79 | |
AK169297.1 | - | 4 - 5 | 892 | 79 | |
BC057133.1 | - | 3 - 4 | 449 | 79 | |
BC057855.1 | - | 3 - 4 | 611 | 79 | |
X78874.1 | - | 3 - 4 | 620 | 79 |