Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdpn
Entrez Gene ID
14726
Gene Name
podoplanin
Gene Aliases
Gp38, OTS-8, RANDAM-2, T1-alpha, T1a, T1alpha
Chromosome Location
Chr.4: 143267409 - 143299564 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010329.3 NP_034459.2 4 - 5 712 67
GenBank mRNA AJ250246.1 - 4 - 5 441 67
AJ297944.2 - 4 - 5 418 67
AK158855.1 - 4 - 5 712 67
BC026551.1 - 4 - 5 596 67
M73748.1 - 4 - 5 569 67
M96645.1 - 4 - 5 479 67

View Full Product Details