The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252479.1 | NP_001239408.1 | 2 - 3 | 504 | 59 |
GenBank mRNA | BC038633.1 | - | 2 - 3 | 343 | 59 |