Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Galntl6
- Entrez Gene ID
-
270049
- Gene Name
- UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 6
- Gene Aliases
-
1700021K10Rik, 4930431L04Rik, A830023L05
- Chromosome Location
-
Chr.8: 57772489 - 58911627 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015274.1 |
- |
3 - 4 |
636 |
86 |
View Full Product Details