Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Auh
Entrez Gene ID
11992
Gene Name
AU RNA binding protein/enoyl-coenzyme A hydratase
Gene Aliases
C77140, W91705
Chromosome Location
Chr.13: 52835102 - 52929706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016709.2 NP_057918.2 2 - 3 293 129
GenBank mRNA AK003219.1 - 2 - 3 272 129
AK003929.1 - 2 - 3 273 129
AK019978.1 - 2 - 3 270 129
AK134886.1 - 2 - 3 265 129
AK147855.1 - 2 - 3 297 129
AK169661.1 - 2 - 3 271 129
BC026525.1 - 2 - 3 231 129
BC049597.1 - 2 - 3 277 129
BY741014.1 - 2 - 3 294 129

View Full Product Details