Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casr
Entrez Gene ID
12374
Gene Name
calcium-sensing receptor
Gene Aliases
CaR, Gprc2a
Chromosome Location
Chr.16: 36490585 - 36562141 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF110179.1 - 4 - 5 1904 60

View Full Product Details