Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ube2ql1
Entrez Gene ID
76980
Gene Name
ubiquitin-conjugating enzyme E2Q family-like 1
Gene Aliases
3110006E14Rik
Chromosome Location
Chr.13: 69702832 - 69739897 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC116979.1 - 1 - 2 389 64
BC116981.1 - 1 - 2 389 64

View Full Product Details