The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033306.1 | 1 - 2 | 200 | 70 | |
GenBank mRNA | AK147147.1 | - | 1 - 2 | 169 | 70 |
BC076501.1 | - | 1 - 2 | 170 | 70 |