Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rusc2
Entrez Gene ID
100213
Gene Name
RUN and SH3 domain containing 2
Gene Aliases
AI840675, mKIAA0375
Chromosome Location
Chr.4: 43381982 - 43427092 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037709.2 NP_001032798.1 7 - 8 3538 59
NM_199057.3 NP_951012.2 7 - 8 3499 59
GenBank mRNA AK053542.1 - 4 - 5 1800 59
AK122263.1 - 7 - 8 3418 59
AK132170.1 - 7 - 8 3428 59
AK166260.1 - 7 - 8 3439 59
BC024790.1 - 5 - 6 729 59
BC141186.1 - 6 - 7 3269 59

View Full Product Details