Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd32
Entrez Gene ID
105377
Gene Name
ankyrin repeat domain 32
Gene Aliases
2700017A04Rik, AW545819, Brctd1, Brctx, C730024G01Rik
Chromosome Location
Chr.13: 77043088 - 77135468 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134071.3 NP_598832.3 2 - 3 222 109
GenBank mRNA AK012253.1 - 2 - 3 215 109
AK047775.1 - 2 - 3 779 109
AK089501.1 - 2 - 3 222 109
AK171473.1 - 2 - 3 222 109
BC061188.1 - 2 - 3 203 109
BC100362.1 - 2 - 3 194 109

View Full Product Details