Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ankrd32
- Entrez Gene ID
-
105377
- Gene Name
- ankyrin repeat domain 32
- Gene Aliases
-
2700017A04Rik, AW545819, Brctd1, Brctx, C730024G01Rik
- Chromosome Location
-
Chr.13: 77043088 - 77135468 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134071.3 |
NP_598832.3 |
9 - 10 |
1260 |
131 |
View Full Product Details