Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samsn1
Entrez Gene ID
67742
Gene Name
SAM domain, SH3 domain and nuclear localization signals, 1
Gene Aliases
4930571B16Rik, 930571B16Rik, Hacs1, SLy2
Chromosome Location
Chr.16: 75858794 - 75909266 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023380.2 NP_075869.2 1 - 2 144 68
GenBank mRNA AK041340.1 - 1 - 2 76 68
AK041566.1 - 1 - 2 119 68
AK053946.1 - 1 - 2 119 68
BC052906.1 - 1 - 2 156 68
BC064778.1 - 1 - 2 144 68
DQ333189.1 - 1 - 2 144 68

View Full Product Details