The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175548.3 | NP_780757.1 | 1 - 2 | 189 | 76 |
GenBank mRNA | AK044845.1 | - | 1 - 2 | 189 | 76 |
AK140043.1 | - | 1 - 2 | 326 | 76 | |
AK158702.1 | - | 1 - 2 | 673 | 76 | |
AK164171.1 | - | - | 496 | 76 |