The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172143.2 | NP_742155.2 | 2 - 3 | 106 | 93 |
GenBank mRNA | AF548110.1 | - | 3 - 4 | 390 | 93 |
AF548111.1 | - | 2 - 3 | 126 | 93 | |
EU683438.1 | - | 2 - 3 | 106 | 93 | |
EU683439.1 | - | 2 - 3 | 106 | 93 |