The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015828.3 | NP_056643.3 | 1 - 2 | 127 | 87 |
GenBank mRNA | AK136573.1 | - | 1 - 2 | 128 | 87 |
AM697710.1 | - | 1 - 2 | 48 | 87 | |
BC015277.1 | - | 1 - 2 | 115 | 87 | |
BC051254.1 | - | 1 - 2 | 115 | 87 |