The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159351.1 | NP_001152823.1 | 1 - 2 | 66 | 80 |
NM_023585.4 | NP_076074.2 | 1 - 2 | 66 | 80 | |
GenBank mRNA | AK033016.1 | - | 1 - 2 | 51 | 80 |
BY025619.1 | - | 1 - 2 | 73 | 80 | |
CJ049747.1 | - | 1 - 2 | 50 | 80 |