Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kazn
Entrez Gene ID
71529
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
2310007B04Rik, 9030409G11Rik, AI256607, AI666764, AW061099, Kaz, Kazrin
Chromosome Location
Chr.4: 142102390 - 142239401 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144531.3 NP_653114.3 10 - 11 1945 80
GenBank mRNA AK032460.1 - 8 - 9 1184 80
AK173090.1 - 10 - 11 1945 80

View Full Product Details