Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbms3
Entrez Gene ID
207181
Gene Name
RNA binding motif, single stranded interacting protein
Gene Aliases
6720477E09Rik, 8430436O14Rik
Chromosome Location
Chr.9: 116572746 - 118040292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172121.1 NP_001165592.1 6 - 7 752 116
NM_001172122.1 NP_001165593.1 5 - 6 1224 116
NM_001172123.1 NP_001165594.1 5 - 6 1224 116
NM_001172124.1 NP_001165595.1 5 - 6 1224 116
NM_001172126.1 NP_001165597.1 5 - 6 1224 116
NM_178660.4 NP_848775.2 5 - 6 1224 116
GenBank mRNA AK030609.1 - 5 - 6 858 116
AK032938.1 - 5 - 6 1221 116
AK033005.1 - 5 - 6 867 116
AK041098.1 - 5 - 6 1226 116
AK046197.1 - 6 - 7 753 116
AK051776.1 - 6 - 7 1271 116
AK052196.1 - 5 - 6 1205 116
BC033604.1 - 1 - 2 79 116
BC108402.1 - 5 - 6 1198 116
BC117841.1 - 5 - 6 664 116

View Full Product Details