Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a34
Entrez Gene ID
384071
Gene Name
solute carrier family 25, member 34
Gene Aliases
Gm1369
Chromosome Location
Chr.4: 141618824 - 141623993 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013780.1 NP_001013802.1 2 - 3 620 73
GenBank mRNA AK142924.1 - 2 - 3 621 73
BC089627.1 - 2 - 3 592 73

View Full Product Details