Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fsd1l
Entrez Gene ID
319636
Gene Name
fibronectin type III and SPRY domain containing 1-like
Gene Aliases
A230072O16Rik, Ccdc10, Csdufd1, Fsd1nl, OTTMUSG00000007002
Chromosome Location
Chr.4: 53629929 - 53707009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195284.1 NP_001182213.1 10 - 11 1153 90
GenBank mRNA BB626514.1 - 3 - 4 280 90

View Full Product Details