The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195284.1 | NP_001182213.1 | 4 - 5 | 477 | 114 |
GenBank mRNA | AK052914.1 | - | 4 - 5 | 520 | 114 |
BB662298.1 | - | 4 - 5 | 515 | 114 |