The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010012.3 | NP_034142.3 | 1 - 1 | 1900 | 77 |
GenBank mRNA | AF090317.1 | - | 1 - 1 | 1886 | 77 |
BC010973.1 | - | 1 - 1 | 1910 | 77 | |
BC010974.1 | - | 1 - 1 | 1903 | 77 | |
BC049969.1 | - | 1 - 1 | 1885 | 77 |