The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133741.2 | NP_598502.2 | 2 - 3 | 191 | 100 |
GenBank mRNA | AK212459.1 | - | 1 - 2 | 57 | 100 |
BC020189.1 | - | 2 - 3 | 82 | 100 |