Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekhm2
Entrez Gene ID
69582
Gene Name
pleckstrin homology domain containing, family M (with RUN domain) member 2
Gene Aliases
2310034J19Rik, AI854247, mKIAA0842
Chromosome Location
Chr.4: 141625732 - 141664151 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033150.1 NP_001028322.1 3 - 4 477 60
GenBank mRNA AK122386.1 - - 561 60
AK219223.1 - 2 - 3 142 60
BC058984.1 - 1 - 2 68 60
CX240883.1 - 3 - 4 477 60

View Full Product Details