Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adamts16
Entrez Gene ID
271127
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 16
Gene Aliases
-
Chromosome Location
Chr.13: 70727798 - 70841848 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172053.2 NP_742050.2 19 - 20 3247 83
GenBank mRNA AK031314.1 - 19 - 20 3378 83
AK173338.1 - 19 - 20 3183 83

View Full Product Details