The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027492.1 | 5 - 6 | 905 | 129 | |
GenBank mRNA | AK030553.1 | - | 5 - 6 | 878 | 129 |
BC094508.1 | - | 5 - 6 | 848 | 129 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | RNA binding motif protein 11 | Rbm11 | A330018F01 | -153.8kb | Anti-sense | Chr. - 75592844 - 75602825 on Build GRCm38 |
Downstream | SAM domain, SH3 domain and nuclear localization signals, 1 | Samsn1 | 4930571B16Rik, 930571B16Rik, Hacs1, SLy2 | 100.1kb | Sense | Chr. - 75858794 - 75909266 on Build GRCm38 |