Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Creb3
Entrez Gene ID
12913
Gene Name
cAMP responsive element binding protein 3
Gene Aliases
AU044960, AW538053, C80076, LZIP, LZIP-1, LZIP-2, Luman
Chromosome Location
Chr.4: 43562634 - 43567060 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013497.3 NP_038525.2 2 - 3 485 82
GenBank mRNA AK090077.1 - 2 - 3 455 82
AK132785.1 - 2 - 3 495 82
AK144208.1 - 2 - 3 488 82
BC002094.1 - 2 - 3 451 82

View Full Product Details